Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Pantolactone 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-(-)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanoneD-(-)-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone(R)-3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-oneD-(-)-Pantoyl Lactone

L15983-06 L15983-14 L15983-14 L15983-06
AAAL15983-14EA 65.19 USD InStock
CAAAAL15983-14 AAAL15983-14 CAAAAL15983-06 AAAL15983-06
D-(-)-Pantolactone 99%
D-(-)-Pantolactone


Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 120…122 °C/15 mm
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00005392
CAS Number: 599-04-2
EINECS: 209-963-3
Order Now
Specification Test Results
Optical rotation -51° (c=3 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty