Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Isocyanato-1-methoxy-4-methylbenzene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methoxy-5-methylphenyl isocyanate

L16001-06 L16001-03 L16001-06 L16001-03
AAAL16001-06EA 70.29 USD InStock
CAAAAL16001-06 AAAL16001-06 CAAAAL16001-03 AAAL16001-03
2-Isocyanato-1-methoxy-4-methylbenzene 97%
2-Isocyanato-1-methoxy-4-methylbenzene


Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 240 °C
Density: 1.112
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013869
CAS Number: 59741-04-7
EINECS: 000-000-0
UN: 2206
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty