Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-γ-Undecalactone 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Undecanoic gamma-Lactone(±)-Undecan-4-olide

L16028-AE L16028-AP L16028-AP L16028-AE
AAAL16028-APEA 105.99 USD InStock
CAAAAL16028-AP AAAL16028-AP CAAAAL16028-AE AAAL16028-AE
(±)-γ-Undecalactone 99%
(±)-γ-Undecalactone


Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 164…166 °C/13 mm
Density: 0.949
Flash Pt: 156 °C(312 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005405
CAS Number: 104-67-6
EINECS: 203-225-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty