Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorodimethylvinylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: DimethylvinylchlorosilaneVinyldimethylchlorosilane

L16198-14 L16198-22 L16198-22 L16198-14
AAAL16198-14EA 64.3 USD InStock
CAAAAL16198-22 CAAAAL16198-14 AAAL16198-14 AAAL16198-22
Chlorodimethylvinylsilane 97%
Chlorodimethylvinylsilane


Formula: C₄H₉ClSi
Boiling Pt: 82…83 °C
Density: 0.883
Flash Pt: -12 °C(10 °F)
Storage Temperature: Ambient
MDL Number: MFCD00018090
CAS Number: 1719-58-0
EINECS: 217-007-1
UN: 2985
ADR: "3",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty