Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(+)-α-Ethylbenzylamine ≥99%, ee 98%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (R)-(+)-1-Phenylpropylamine(R)-1-Phenylpropan-1-amine

L16319-14 L16319-06 L16319-06 L16319-14
AAAL16319-14EA 268.99 USD InStock
CAAAAL16319-14 AAAL16319-14 CAAAAL16319-06 AAAL16319-06
(R)-(+)-α-Ethylbenzylamine ≥99%, ee 98%, ChiPros®
(R)-(+)-α-Ethylbenzylamine
For synthesis of optically active products


Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 205 °C
Melting Pt: –69 °C
Density: 0.930
Flash Pt: 77 °C(170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00083057
CAS Number: 3082-64-2
EINECS: 608-544-6
UN: 2735
ADR: "8",III
Order Now
Specification Test Results
Optical rotation +20° (neat)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty