Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodoperfluoropropane 97% stabilized, powder

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-IodoheptafluoropropaneHeptafluoro-2-iodopropane1,1,1,2,3,3,3-Heptafluoro-2-iodopropaneHeptafluoroisopropyl iodide

Stabilization: Stabilised with copper

L16523-14 L16523-06 L16523-22 L16523-22 L16523-14 L16523-06
AAAL16523-22EA 344.99 USD InStock
AAAL16523-22 CAAAAL16523-22 AAAL16523-06 AAAL16523-14 CAAAAL16523-06 CAAAAL16523-14
2-Iodoperfluoropropane 97% stabilized, powder
2-Iodoperfluoropropane


Formula: C₃F₇I
MW: 295.93 g/mol
Boiling Pt: 38 °C
Melting Pt: –56 °C
Density: 2.100
Storage Temperature: Ambient
MDL Number: MFCD00042089
CAS Number: 677-69-0
EINECS: 211-643-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty