Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1H,1H,9H,9H-Perfluoro-1,9-nonanediol tech. 90%

Supplier: Thermo Scientific Chemicals
Synonyms: 1H,1H,9H,9H-Perfluoro-1,9-nonanediol2,2,3,3,4,4,5,5,6,6,7,7,8,8-Tetradecafluoro-1,9-nonanediol2,2,3,3,4,4,5,5,6,6,7,7,8,8-Tetradecafluoro-1,9-nonandiol

L16599-14 L16599-14 L16599-06 L16599-06
AAAL16599-14EA 196.99 USD InStock
AAAL16599-14 CAAAAL16599-06 CAAAAL16599-14 AAAL16599-06
1H,1H,9H,9H-Perfluoro-1,9-nonanediol tech. 90%
1H,1H,9H,9H-Tetradecafluoro-1,9-nonandiol


Formula: C₉H₆F₁₄O₂
MW: 412.12 g/mol
Boiling Pt: 140 °C/5 mm
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00153275
CAS Number: 203303-01-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty