Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,3,5-Tris(3,3,3-trifluoropropyl)-1,3,5-trimethylcyclotrisiloxane1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane

L16680-06 L16680-14 L16680-22 L16680-22 L16680-06 L16680-14
AAAL16680-22EA 545.99 USD InStock
AAAL16680-22 CAAAAL16680-22 AAAL16680-06 AAAL16680-14 CAAAAL16680-06 CAAAAL16680-14
1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane 97%
1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane


Formula: C₁₂H₂₁F₉O₃Si₃
MW: 468.54 g/mol
Boiling Pt: 135 °C/22 mm
Melting Pt: 34…36 °C
Density: 1.240
Storage Temperature: Ambient
MDL Number: MFCD00054667
CAS Number: 2374-14-3
EINECS: 219-154-7
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty