Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultramark® 1621, Ultramark® mass spec standard

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Massmark 1621

L16698-MD L16698-MD L16698-03 L16698-03
AAAL16698-03EA 133.79 USD InStock
CAAAAL16698-03 AAAL16698-03 CAAAAL16698-MD AAAL16698-MD
Ultramark® 1621, Ultramark® mass spec standard
Ultramark® 1621


Formula: C₆H₁₈N₃P₃
MW: 231.189 g/mol
Density: 1.8
Storage Temperature: Ambient
MDL Number: MFCD00242454
CAS Number: 105809-15-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty