Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(2,2,2-trifluoroethyl)amine 95%

Supplier: Thermo Scientific Chemicals
Danger

L16709-03 L16709-06 L16709-03 L16709-06
AAAL16709-06EA 239.99 USD InStock
AAAL16709-06 AAAL16709-03 CAAAAL16709-06 CAAAAL16709-03
Bis(2,2,2-trifluoroethyl)amine 95%
Bis(2,2,2-trifluoroethyl)amine


Formula: C₄H₅F₆N
Boiling Pt: 70…71 °C
Density: 1.100
Storage Temperature: Ambient
EINECS: 206-978-7
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty