Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Heptafluorobutyro)phenone 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,2,3,3,4,4,4-Heptafluorobutyrophenone

L16777-06 L16777-22 L16777-22 L16777-06 L16777-14 L16777-14
AAAL16777-14EA 311.99 USD InStock
AAAL16777-14 CAAAAL16777-06 AAAL16777-06 CAAAAL16777-22 AAAL16777-22 CAAAAL16777-14
(Heptafluorobutyro)phenone 99%
(Heptafluorobutyro)phenone


Formula: C₁₀H₅F₇O
MW: 274.14
Boiling Pt: 170…172 °C
Density: 1.473
Storage Temperature: Ambient
MDL Number: MFCD00077552
CAS Number: 559-91-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty