Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,3,3,3-Pentafluoropropylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

L16819-03 L16819-06 L16819-06 L16819-03
AAAL16819-03EA 111.99 USD InStock
CAAAAL16819-03 AAAL16819-03 AAAL16819-06 CAAAAL16819-06
2,2,3,3,3-Pentafluoropropylamine 97%
2,2,3,3,3-Pentafluoropropylamine


Formula: C₃H₄F₅N
Boiling Pt: 49…50 °C
Density: 1.400
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042459
CAS Number: 422-03-7
EINECS: 207-010-6
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty