Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine 90%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Chloro-N,N-diethyl-1,1,2-trifluoroethylamine

L17330-14 L17330-06 L17330-22 L17330-22 L17330-14 L17330-06
AAAL17330-22EA 480.99 USD InStock
AAAL17330-22 CAAAAL17330-06 AAAL17330-06 CAAAAL17330-22 AAAL17330-14 CAAAAL17330-14
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine 90%
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine


Formula: C₆H₁₁ClF₃N
MW: 189.61 g/mol
Boiling Pt: 38 °C/10 mm
Density: 1.190
Storage Temperature: Ambient
MDL Number: MFCD00054672
CAS Number: 357-83-5
EINECS: 000-000-0
UN: 3267
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty