Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perfluorononane 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Eicosafluorononane

L17363-14 L17363-06 L17363-14 L17363-06
AAAL17363-14EA 127.99 USD InStock
CAAAAL17363-14 CAAAAL17363-06 AAAL17363-14 AAAL17363-06
Perfluorononane 99%
Perfluorononane


Formula: C₉F₂₀
Boiling Pt: 125…126 °C
Melting Pt: –16 °C
Density: 1,799
Storage Temperature: Ambient
MDL Number: MFCD00153254
CAS Number: 375-96-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty