Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-3-(tert-butyldimethylsiloxy)benzene ≥98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (3-Bromophenoxy)(tert-butyl)dimethylsilane

L17440-06 L17440-06 L17440-03 L17440-03
AAAL17440-03EA 40.12 USD InStock
CAAAAL17440-06 AAAL17440-03 AAAL17440-06 CAAAAL17440-03
1-Bromo-3-(tert-butyldimethylsiloxy)benzene ≥98%
1-Bromo-3-(tert-butyldimethylsiloxy)benzene


Formula: C₁₂H₁₉BrOSi
MW: 287.27 g/mol
Boiling Pt: 66 °C/0.3 mm
Storage Temperature: Ambient
MDL Number: MFCD01318106
CAS Number: 65423-56-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty