Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Isopropylphenylboronic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Isopropylbenzeneboronic acid

L17459-03 L17459-06 L17459-06 L17459-03
AAAL17459-06EA 427.99 USD InStock
CAAAAL17459-03 AAAL17459-06 AAAL17459-03 CAAAAL17459-06
4-Isopropylphenylboronic acid 98+%
4-Isopropylphenylboronic acid


Formula: C₉H₁₃BO₂
MW: 164.01 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD01074614
CAS Number: 16152-51-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty