Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Methylindole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Methyl-1H-indole

L17509-03 L17509-03 L17509-MD L17509-MD
AAAL17509-03EA 180.99 USD InStock
AAAL17509-03 CAAAAL17509-03 AAAL17509-MD CAAAAL17509-MD
6-Methylindole 98%
6-Methylindole


Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 112 °C/5 mm
Melting Pt: 29…32 °C
Density: 1.059
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005682
CAS Number: 3420-02-8
ADR: 9,
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty