Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methyl-2-hexanone 99%

Supplier: Thermo Scientific Chemicals
Warning

L17584-AE L17584-AP L17584-0F
AAAL17584-AEEA 26.2 USD InStock
AAAL17584-AE AAAL17584-0F AAAL17584-AP
5-Methyl-2-hexanone 99%
5-Methyl-2-hexanone


Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 144…145 °C
Melting Pt: –74 °C
Density: 0.812
Flash Pt: 41 °C(105 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008950
CAS Number: 110-12-3
EINECS: 203-737-8
UN: 2302
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty