Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-indole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-Butyl 1-indolecarboxylate

L17601-06 L17601-03 L17601-03 L17601-06
AAAL17601-06EA 153.99 USD InStock
AAAL17601-06 CAAAAL17601-03 AAAL17601-03 CAAAAL17601-06
1-Boc-indole 97%
1-Boc-indole


Formula: C₁₃H₁₅NO₂
MW: 217.27 g/mol
Boiling Pt: 113…114 °C/0.2 mm
Density: 1.070
Storage Temperature: Ambient
MDL Number: MFCD02093939
CAS Number: 75400-67-8
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty