Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Isopropenyl-2-benzimidazolidinone ≥98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-(Prop-1-en-2-yl)-1H-benzo[d]imidazol-2(3H)-one1-(Prop-1-en-2-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one

L17626-03 L17626-06 L17626-03 L17626-06
AAAL17626-03EA 40.59 USD InStock
AAAL17626-03 CAAAAL17626-03 AAAL17626-06 CAAAAL17626-06
1-Isopropenyl-2-benzimidazolidinone ≥98%
1-Isopropenyl-2-benzimidazolidinone


Formula: C₁₀H₁₀N₂O
MW: 174.203 g/mol
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00218253
CAS Number: 52099-72-6
EINECS: 257-661-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty