Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-(tert-Butoxycarbonylamino)-1-pentanol 96%

Supplier: Thermo Scientific Chemicals
Synonyms: tert-Butyl (5-hydroxypentyl)carbamateN-(5-Hydroxypentyl)carbamic Acid tert-Butyl EsterN-Boc-5-aminopentanoltert-Butyl N-(5-Hydroxypentyl)carbamate

L17649-06 L17649-03 L17649-06 L17649-03
AAAL17649-03EA 62.89 USD InStock
CAAAAL17649-03 AAAL17649-03 CAAAAL17649-06 AAAL17649-06
5-(tert-Butoxycarbonylamino)-1-pentanol 96%
5-(tert-Butoxycarbonylamino)-1-pentanol


Formula: C₁₀H₂₁NO₃
MW: 203.28 g/mol
Density: 1.000
Flash Pt: >109 °C(228 °F)
Storage Temperature: Ambient
MDL Number: MFCD01862955
CAS Number: 75178-90-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty