Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-bromo-3,4-dimethylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

L17672-06 L17672-06 L17672-03 L17672-03
AAAL17672-06EA 315.99 USD InStock
AAAL17672-06 CAAAAL17672-06 AAAL17672-03 CAAAAL17672-03
2-Amino-5-bromo-3,4-dimethylpyridine 97%
2-Amino-5-bromo-3,4-dimethylpyridine


Formula: C₇H₉BrN₂
MW: 201.07 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD02093947
CAS Number: 374537-97-0
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty