Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Bromo-1-hydroxyisoquinoline ≥98%

Supplier: Thermo Scientific Chemicals
Warning

L17800-06 L17800-03 L17800-06 L17800-03
AAAL17800-06EA 371.99 USD InStock
AAAL17800-06 CAAAAL17800-06 AAAL17800-03 CAAAAL17800-03
7-Bromo-1-hydroxyisoquinoline ≥98%
7-Bromo-1-hydroxyisoquinoline


Formula: C₉H₆BrNO
MW: 224.06 g/mol
Melting Pt: 248…250 °C
Storage Temperature: Ambient
MDL Number: MFCD02093963
CAS Number: 223671-15-6
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty