Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chlorosalicylamide 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Chloro-2-hydroxybenzamide

L17897-06 L17897-06 L17897-14 L17897-14
AAAL17897-06EA 63.59 USD InStock
AAAL17897-06 AAAL17897-14 CAAAAL17897-14 CAAAAL17897-06
5-Chlorosalicylamide 97%
5-Chlorosalicylamide


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 223…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00007980
CAS Number: 7120-43-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty