Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-indole-2-boronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acidN-Boc-indole-2-boronic acid1H-Indole-2-boronic acid, N-BOC protectedN-Boc-indole-2-boronic Acid1-(tert-Butoxycarbonyl)indole-2-boronic acid

L18009-03 L18009-MD L18009-06 L18009-03 L18009-MD L18009-06
AAAL18009-03EA 108.99 USD InStock
CAAAAL18009-03 AAAL18009-03 CAAAAL18009-MD CAAAAL18009-06 AAAL18009-MD AAAL18009-06
1-Boc-indole-2-boronic acid 95%
1-Boc-indole-2-boronic acid


Formula: C₁₃H₁₆BNO₄
MW: 261.08 g/mol
Melting Pt: ca 85 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02093045
CAS Number: 213318-44-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty