Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Bromomethyl)phenoxyacetic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

L18029-03 L18029-03 L18029-06 L18029-06
AAAL18029-06EA 149.99 USD InStock
CAAAAL18029-06 AAAL18029-06 CAAAAL18029-03 AAAL18029-03
3-(Bromomethyl)phenoxyacetic acid 97%
3-(Bromomethyl)phenoxyacetic acid


Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD02093985
CAS Number: 136645-25-5
EINECS: 000-000-0
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty