Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Cyclohexylbenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L18060-06 L18060-03 L18060-03 L18060-06
AAAL18060-03EA 97.69 USD InStock
CAAAAL18060-03 AAAL18060-03 AAAL18060-06 CAAAAL18060-06
2-Cyclohexylbenzoic acid 97%
2-Cyclohexylbenzoic acid


Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00452653
CAS Number: 97023-48-8
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty