Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminophenylboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2-Aminophenyl)metaboric acid(2-Aminophenyl)boronic acidDihydroxy(2-aminophenyl)borane2-Aminobenzeneboronic acid

L18069-06 L18069-06 L18069-MC L18069-MC L18069-03 L18069-03
AAAL18069-06EA 648.99 USD InStock
CAAAAL18069-06 CAAAAL18069-MC AAAL18069-06 AAAL18069-MC AAAL18069-03 CAAAAL18069-03
2-Aminophenylboronic acid 96%
2-Aminophenylboronic acid


Formula: C₆H₈BNO₂
MW: 136.95 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD01074645
CAS Number: 5570-18-3
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty