Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-o-toluic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloro-6-methylbenzoic acid

L18348-03 L18348-03 L18348-06 L18348-06
AAAL18348-06EA 269.99 USD InStock
CAAAAL18348-03 CAAAAL18348-06 AAAL18348-06 AAAL18348-03
6-Chloro-o-toluic acid 97%
6-Chloro-o-toluic acid


Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00045799
CAS Number: 21327-86-6
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty