Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Dimethylamino)benzoyl chloride hydrochloride tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

L18433-03 L18433-03 L18433-06 L18433-06
AAAL18433-03EA 83.69 USD InStock
CAAAAL18433-03 AAAL18433-03 CAAAAL18433-06 AAAL18433-06
3-(Dimethylamino)benzoyl chloride hydrochloride tech. 90%, Technical Grade
3-(Dimethylamino)benzoyl chloride hydrochloride


Formula: C₉H₁₁Cl₂NO
MW: 220.1 g/mol
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD19442144
CAS Number: 117500-61-5
EINECS: 000-000-0
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty