Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-D-2-phenylglycine 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Boc-D-2-phenylglycineBOC-D-alpha-phenylglycineBoc-D-Phg-OH

L18540-06 L18540-06 L18540-03 L18540-03
AAAL18540-06EA 151.99 USD InStock
CAAAAL18540-03 AAAL18540-06 CAAAAL18540-06 AAAL18540-03
N-(tert-Butoxycarbonyl)-D-2-phenylglycine 99%
N-(tert-Butoxycarbonyl)-D-2-phenylglycine


Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00062043
CAS Number: 33125-05-2
Order Now
Specification Test Results
Optical rotation -144° (c=1 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty