Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-3,4-dimethylisoxazole 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 3,4-Dimethylisoxazol-5-amine3,4-Dimethylisoxazol-5-ylamine

L18668-06 L18668-06 L18668-03 L18668-03
AAAL18668-03EA 44.69 USD InStock
AAAL18668-03 CAAAAL18668-06 CAAAAL18668-03 AAAL18668-06
5-Amino-3,4-dimethylisoxazole 99%
5-Amino-3,4-dimethylisoxazole


Formula: C₅H₈N₂O
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00046081
CAS Number: 19947-75-2
EINECS: 243-437-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty