Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allyl isocyanate 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Isocyanatoprop-1-ene

L18710-06 L18710-06 L18710-03 L18710-03
AAAL18710-03EA 36.19 USD InStock
CAAAAL18710-06 AAAL18710-03 AAAL18710-06 CAAAAL18710-03
Allyl isocyanate 96%
Allyl isocyanate


Formula: C₄H₅NO
MW: 83.09 g/mol
Boiling Pt: 87…89 °C
Density: 0.940
Flash Pt: 43 °C(109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002044
CAS Number: 1476-23-9
EINECS: 000-000-0
UN: 2478
ADR: "3",III
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty