Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-m-anisic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Methoxy-2-methylbenzoic acid

L18729-18 L18729-09 L18729-09 L18729-18
AAAL18729-09EA 57.49 USD InStock
AAAL18729-09 CAAAAL18729-09 AAAL18729-18 CAAAAL18729-18
2-Methyl-m-anisic acid 97%
2-Methyl-m-anisic acid


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094039
CAS Number: 55289-06-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty