Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-4-Methyl-2-pentanol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-(+)-Methyl isobutyl carbinol

L18882-03 L18882-03 L18882-MD L18882-MD
AAAL18882-MDEA 69.19 USD InStock
AAAL18882-MD AAAL18882-03 CAAAAL18882-MD CAAAAL18882-03
(S)-(+)-4-Methyl-2-pentanol 99%
(S)-(+)-4-Methyl-2-pentanol
For synthesis of optically active products


Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 130…131 °C
Density: 0.802
Flash Pt: 41 °C(105 °F)
Storage Temperature: Ambient
MDL Number: MFCD03093078
CAS Number: 14898-80-7
UN: 2053
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty