Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methylnicotinamide 97%

Supplier: Thermo Scientific Chemicals
Warning

L18970-06 L18970-03 L18970-06 L18970-03
AAAL18970-06EA 257.99 USD InStock
CAAAAL18970-03 AAAL18970-06 CAAAAL18970-06 AAAL18970-03
5-Methylnicotinamide 97%
5-Methylnicotinamide


Formula: C₇H₈N₂O
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD05664041
CAS Number: 70-57-5
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty