Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(-)-3-Hydroxy-3-phenylpropionic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-3-Hydroxy-3-phenylpropanoic acid

L19041-03 L19041-MD L19041-MD L19041-03
AAAL19041-MDEA 74.69 USD InStock
CAAAAL19041-MD AAAL19041-MD CAAAAL19041-03 AAAL19041-03
(S)-(-)-3-Hydroxy-3-phenylpropionic acid 98+%
(S)-(-)-3-Hydroxy-3-phenylpropionic acid


Formula: C₉H₁₀O₃
MW: 166.18
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00145262
CAS Number: 36567-72-3
Order Now
Specification Test Results
Optical rotation -22° (c=4 in methanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty