Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methoxyindol-3-ylacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 5-Methoxy-3-indoleacetic acid5-Methoxyindole-3-acetic acid2-(5-Methoxy-1H-indol-3-yl)acetic acid

L19073-03 L19073-03 L19073-MD L19073-MD
AAAL19073-03EA 135.99 USD InStock
AAAL19073-03 CAAAAL19073-03 AAAL19073-MD CAAAAL19073-MD
5-Methoxyindol-3-ylacetic acid 98+%
5-Methoxyindol-3-ylacetic acid


Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
Melting Pt: 145…148 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005638
CAS Number: 3471-31-6
EINECS: 222-438-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty