Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(+)-4-Chloro-α-methylbenzylamine 97%, ee 98%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (R)-1-(4-Chlorophenyl)ethanamine(R)-1-(4-Chlorophenyl)ethylamine(R)-(+)-1-(4-Chlorophenyl)ethylamine

L19119-06 L19119-06 L19119-03 L19119-03 L19119-14
AAAL19119-03EA 46.49 USD InStock
AAAL19119-03 AAAL19119-14 CAAAAL19119-06 CAAAAL19119-03 AAAL19119-06
(R)-(+)-4-Chloro-α-methylbenzylamine 97%, ee 98%, ChiPros®
(R)-(+)-4-Chloro-α-methylbenzylamine


Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 232 °C
Density: 1.110
Flash Pt: 115 °C(239 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671639
CAS Number: 27298-99-3
UN: 2810
ADR: "6.1",III
Order Now
Specification Test Results
Optical rotation +31° (neat)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty