Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-5-nitrobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

L19233-03 L19233-06 L19233-06 L19233-03
AAAL19233-03EA 77.79 USD InStock
CAAAAL19233-03 AAAL19233-03 CAAAAL19233-06 AAAL19233-06
2-Fluoro-5-nitrobenzaldehyde 98%
2-Fluoro-5-nitrobenzaldehyde


Formula: C₇H₄FNO₃
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00042298
CAS Number: 27996-87-8
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty