Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethynyl-benzoic acid sodium salt 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium 4-ethynylbenzoate

L19292-06 L19292-03 L19292-MD L19292-06 L19292-03
AAAL19292-03EA 99.49 USD InStock
CAAAAL19292-06 AAAL19292-03 CAAAAL19292-MD AAAL19292-06 CAAAAL19292-03
4-Ethynyl-benzoic acid sodium salt 97%
4-Ethynyl-benzoic acid sodium salt


Formula: C₉H₅NaO₂
MW: 168.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426026
CAS Number: 144693-65-2
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty