Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(+)-1-Phenylethanol ≥99%, ee 97%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (R)-(+)-α-Methylbenzyl alcohol(R)-(+)-sec-Phenethyl alcohol(R)-1-Phenylethanol

L19296-06 L19296-14 L19296-06 L19296-14
AAAL19296-06EA 126.8 USD InStock
AAAL19296-06 AAAL19296-14 CAAAAL19296-06 CAAAAL19296-14
(R)-(+)-1-Phenylethanol ≥99%, ee 97%, ChiPros®
(R)-(+)-1-Phenylethanol
For synthesis of optically active products


Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 88…89 °C/10 mm
Melting Pt: 9…11 °C
Density: 1.012
Flash Pt: 85 °C(185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064263
CAS Number: 1517-69-7
EINECS: 604-814-2
UN: 2937
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty