Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Fluoro-2-hydroxybenzaldehyde

L19324-06 L19324-03 L19324-MD L19324-06 L19324-MD L19324-03
AAAL19324-03EA 143.99 USD InStock
CAAAAL19324-MD AAAL19324-03 CAAAAL19324-03 AAAL19324-MD AAAL19324-06 CAAAAL19324-06
3-Fluorosalicylaldehyde 98%
3-Fluorosalicylaldehyde


Formula: C₇H₅FO₂
MW: 140.12 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00003319
CAS Number: 394-50-3
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty