Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-ethylenediamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: tert-Butyl (2-aminoethyl)carbamate hydrochloride

L19485-03 L19485-06 L19485-03 L19485-06
AAAL19485-03EA 109.99 USD InStock
CAAAAL19485-06 CAAAAL19485-03 AAAL19485-03 AAAL19485-06
N-Boc-ethylenediamine hydrochloride 98%
N-Boc-ethylenediamine hydrochloride


Formula: C₇H₁₇ClN₂O₂
MW: 196.68 g/mol
Melting Pt: 148…151 °C
Storage Temperature: Ambient
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty