Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-1,3-propanediamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-Butyl (3-aminopropyl)carbamate hydrochloride

L19486-03 L19486-MD L19486-MD L19486-03
AAAL19486-03EA 114.99 USD InStock
CAAAAL19486-03 CAAAAL19486-MD AAAL19486-03 AAAL19486-MD
N-Boc-1,3-propanediamine hydrochloride 98%
N-Boc-1,3-propanediamine hydrochloride


Formula: C₈H₁₉ClN₂O₂
MW: 210.702 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD01076598
CAS Number: 127346-48-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty