Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-1,6-hexanediamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Boc-1,6-diaminohexane·HClN-Boc-1,6-diaminohexane hydrochlorideN-1-Boc-1,6-diaminohexane·HCl

L19487-06 L19487-03 L19487-06 L19487-03
AAAL19487-06EA 206.99 USD InStock
CAAAAL19487-03 AAAL19487-06 CAAAAL19487-06 AAAL19487-03
N-Boc-1,6-hexanediamine hydrochloride 98%
N-Boc-1,6-hexanediamine hydrochloride


Formula: C₁₁H₂₄N₂O₂·HCl
MW: 252.783 g/mol
Melting Pt: ca 164 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00039072
CAS Number: 65915-94-8
EINECS: 265-976-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty