Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-formylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Bromopyridine-2-carboxaldehyde6-Bromopyridine-2-carbaldehyde6-Bromo-2-pyridinecarboxaldehyde

L19518-06 L19518-03 L19518-03 L19518-06 L19518-14 L19518-14
AAAL19518-06EA 143.99 USD InStock
CAAAAL19518-06 AAAL19518-06 CAAAAL19518-03 CAAAAL19518-14 AAAL19518-03 AAAL19518-14
2-Bromo-6-formylpyridine 97%
2-Bromo-6-formylpyridine


Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD02683546
CAS Number: 34160-40-2
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty