Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Benzo[b]furylacetonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Benzofuran-3-ylacetonitrile

L19550-03 L19550-MD L19550-03 L19550-MD
AAAL19550-03EA 226.99 USD InStock
AAAL19550-03 CAAAAL19550-03 AAAL19550-MD CAAAAL19550-MD
3-Benzo[b]furylacetonitrile 99%
3-Benzo[b]furylacetonitrile


Formula: C₁₀H₇NO
MW: 157.17 g/mol
Boiling Pt: 120…122 °C/4 mm
Melting Pt: 38…39 °C
Storage Temperature: Ambient
MDL Number: MFCD02183555
CAS Number: 52407-43-9
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty