Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylphenylboronic acid neopentyl glycol ester 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 5,5-Dimethyl-2-(p-tolyl)-1,3,2-dioxaborinane4-Methylphenylboronic acid neopentyl ester4-Methylbenzeneboronic acid neopentyl glycol ester

L19563-03 L19563-03 L19563-14 L19563-06 L19563-06
AAAL19563-03EA 38.39 USD InStock
AAAL19563-03 CAAAAL19563-06 AAAL19563-06 CAAAAL19563-03 CAAAAL19563-14
4-Methylphenylboronic acid neopentyl glycol ester 99%
4-Methylphenylboronic acid neopentyl glycol ester


Formula: C₁₂H₁₇BO₂
MW: 204.08 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD03788728
CAS Number: 380481-66-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty