Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromopyridine-2-carbonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Bromopicolinonitrile5-Bromo-2-cyanopyridine

L19621-03 L19621-06 L19621-06 L19621-03
AAAL19621-03EA 52.69 USD InStock
CAAAAL19621-03 CAAAAL19621-06 AAAL19621-03 AAAL19621-06
5-Bromopyridine-2-carbonitrile 95%
5-Bromopyridine-2-carbonitrile


Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Melting Pt: 124…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00234144
CAS Number: 97483-77-7
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty